Learn More
National Safety Apparel™ Enespro™ 16 cal FR Balaclava
SDP
Catalog No. 17355205 Shop All National Safety Apparel Products
Encompass_Preferred
This item is not returnable. View return policy
Catalog No. 17-355-205 Supplier National Safety Apparel™ Supplier No. H11RY
This item is not returnable. View return policy
FR balaclava is durable, lightweight, and designed for comfort and flexibility.
  • Flat lock seam for comfort when worn under a hard hat
  • Single layer top and bib
  • Large face opening
  • Durable and lightweight
  • FR fabric will not melt, drip, or ignite
Specifications
Closure Type Pull-over
Disposable Reusable
Arc Flash Rating 16 cal/cm²
Certifications/Compliance ASTM F1506, OSHA 1910.269, NFPA 70E, CSA Z462
Quantity 1 Ea.
Weight (English) 6.5 oz.
Includes Balaclava
Construction Balaclava
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Resistance Flame
Hood Type Attached
Style Balaclava