Learn More
Cayman Chemical LIgnocerIc AcId 100mg
SDP
Catalog No. 502954993
Encompass_Preferred
A 24-carbon saturated (24:0) fatty acid that is synthesized during brain development and is found in cerebrosides
Catalog No. 50-295-4993 Supplier Cayman Chemical Supplier No. 13353100 MG
A 24-carbon saturated (24:0) fatty acid that is synthesized during brain development and is found in cerebrosides