Learn More
Cayman Chemical LIgnocerIc AcId 50mg
SDP
Catalog No. 502953514
Encompass_Preferred
A 24-carbon saturated (24:0) fatty acid that is synthesized during brain development and is found in cerebrosides
Catalog No. 50-295-3514 Supplier Cayman Chemical Supplier No. 1335350 MG
A 24-carbon saturated (24:0) fatty acid that is synthesized during brain development and is found in cerebrosides