Learn More
Medchemexpress LLC Lignoceric acid | 557-59-5 | 98.0% | 100 MG
SDP
Catalog No. 5000230896
Encompass_Preferred
Lignoceric acid (tetracosanoic acid) is a 24-carbon saturated (24:0) long-chain fatty acid supplied as a solid for laboratory use. It is used as a reagent and reference standard in lipid research and analytical chemistry.
  • 24-carbon saturated fatty acid (C24:0).
  • Molecular weight 368.64 g/mol; formula C24H48O2.
  • High purity, listed as 98.0%.
  • Available in small pack sizes suitable for analytical work, including 100 mg.
  • Solid, white to off-white appearance.
Catalog No. 50-002-30896 Supplier Medchemexpress LLC Supplier No. HY121883100MG
Lignoceric acid (tetracosanoic acid) is a 24-carbon saturated (24:0) long-chain fatty acid supplied as a solid for laboratory use. It is used as a reagent and reference standard in lipid research and analytical chemistry.
  • 24-carbon saturated fatty acid (C24:0).
  • Molecular weight 368.64 g/mol; formula C24H48O2.
  • High purity, listed as 98.0%.
  • Available in small pack sizes suitable for analytical work, including 100 mg.
  • Solid, white to off-white appearance.