Learn More
Medchemexpress LLC Lignoceric acid | 557-59-5 | 98.0% | 50 MG
SDP
Catalog No. 5000231040
Encompass_Preferred
Lignoceric acid is a long-chain, saturated fatty acid (tetracosanoic acid, C24:0) used as a reagent and reference standard in lipid research and metabolic studies.
  • Long-chain saturated fatty acid (C24:0)
  • Molecular formula C24H48O2, molecular weight 368.64 g/mol
  • White to off-white solid appearance
  • Commonly used in lipid analysis and metabolic studies
  • Available in small research pack sizes such as 50 mg
Catalog No. 50-002-31040 Supplier Medchemexpress LLC Supplier No. HY12188350MG
Add to Cart
Add to Cart
Lignoceric acid is a long-chain, saturated fatty acid (tetracosanoic acid, C24:0) used as a reagent and reference standard in lipid research and metabolic studies.
  • Long-chain saturated fatty acid (C24:0)
  • Molecular formula C24H48O2, molecular weight 368.64 g/mol
  • White to off-white solid appearance
  • Commonly used in lipid analysis and metabolic studies
  • Available in small research pack sizes such as 50 mg