Learn More
Cayman Chemical Lignoceric Acid-d47, Purity: ≥99% deuterated forms (d1-d47), CAS: 68060-00-4, 25 mg
SDP
Catalog No. 502892918
Encompass_Preferred
An internal standard for the quantification of lignoceric acid by GC- or LC-MS
Catalog No. 50-289-2918 Supplier Cayman Chemical Supplier No. 900332025 MG
An internal standard for the quantification of lignoceric acid by GC- or LC-MS