Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lignoceric acid-d9 | 2415226-90-1 | 98.0% | 10 MG
SDP

Catalog No. 5000231221
Encompass_Preferred

Lignoceric acid-d9 is the deuterium-labeled (d9) analogue of lignoceric (tetracosanoic) acid used as a stable isotope internal standard in analytical and metabolic studies. Supplied as a white to off-white solid with high isotopic purity, it is provided in small packaged quantities for use in mass spectrometry, tracer experiments, and quantitative lipid analysis.

  • Deuterium-labeled (d9) stable isotope standard for mass spectrometry.
  • Molecular formula C24H39D9O2; molecular weight 377.69.
  • CAS number 2415226-90-1 for substance identification.
  • High purity, typically 98.0%.
  • Available in small packaged sizes suitable for analytical use (e.g., 10 mg).
  • White to off-white solid; handle according to SDS instructions.

Catalog No. 50-002-31221 Supplier Medchemexpress LLC Supplier No. HY121883SS510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lignoceric acid-d9 is the deuterium-labeled (d9) analogue of lignoceric (tetracosanoic) acid used as a stable isotope internal standard in analytical and metabolic studies. Supplied as a white to off-white solid with high isotopic purity, it is provided in small packaged quantities for use in mass spectrometry, tracer experiments, and quantitative lipid analysis.

  • Deuterium-labeled (d9) stable isotope standard for mass spectrometry.
  • Molecular formula C24H39D9O2; molecular weight 377.69.
  • CAS number 2415226-90-1 for substance identification.
  • High purity, typically 98.0%.
  • Available in small packaged sizes suitable for analytical use (e.g., 10 mg).
  • White to off-white solid; handle according to SDS instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.