Learn More
Cayman Chemical Lignoceroyl Ethanolamide, Purity: ≥98%, CAS: 10015-68-6, 10 mg
SDP
Catalog No. 502893098
Encompass_Preferred
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids
Catalog No. 50-289-3098 Supplier Cayman Chemical Supplier No. 900174410 MG
A member of the family of fatty N-acyl ethanolamines collectively called endocannabinoids