Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside | 199796-12-8 | 1 MG
SDP

Catalog No. 5000421150
Encompass_Preferred

Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside is a flavanone glycoside isolated from Glycyrrhizia inflata. It is intended for research use only.

  • Purity: 99.56%
  • Appearance: White to yellow solid
  • Molecular formula: C26H30O13
  • Molecular weight: 550.51
  • Recommended storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-21150 Supplier Medchemexpress LLC Supplier No. HYN26241MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside is a flavanone glycoside isolated from Glycyrrhizia inflata. It is intended for research use only.

  • Purity: 99.56%
  • Appearance: White to yellow solid
  • Molecular formula: C26H30O13
  • Molecular weight: 550.51
  • Recommended storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.