Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside | 199796-12-8 | 99.6% | 550.51 g·mol⁻¹ | C26H30O13 | 10 MG
SDP

Catalog No. 5000288975
Encompass_Preferred

Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside is a flavanone glycoside isolated from Glycyrrhizia inflata and provided as a solid research reagent for analytical and biochemical studies.

  • Flavanone glycoside isolated from Glycyrrhizia inflata.
  • Molecular formula C26H30O13.
  • Molecular weight 550.51 g·mol⁻¹.
  • CAS number 199796-12-8.
  • Purity 99.6% by supplier analysis.
  • Physical form solid, supplied in small milligram quantities.
  • Storage: 4°C protected from light; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-88975 Supplier Medchemexpress LLC Supplier No. HYN262410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Liguiritigenin-7-O-D-apiosyl-4'-O-D-glucoside is a flavanone glycoside isolated from Glycyrrhizia inflata and provided as a solid research reagent for analytical and biochemical studies.

  • Flavanone glycoside isolated from Glycyrrhizia inflata.
  • Molecular formula C26H30O13.
  • Molecular weight 550.51 g·mol⁻¹.
  • CAS number 199796-12-8.
  • Purity 99.6% by supplier analysis.
  • Physical form solid, supplied in small milligram quantities.
  • Storage: 4°C protected from light; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.