Learn More
Lkt Laboratories Ligustrazine Hydrochloride, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories L33261G
Encompass_Preferred
Dihydropyrazine found in Ligusticum walliichi. PDE10A2 inhibitor, adenosine 2A/2B antagonist. CAS: 76494-51-4. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-5019