Learn More
Selleck Chemical LLC Ligustrazine hydrochloride S3775-25mg
SDP
Catalog No. 503109635
Encompass_Preferred
Ligustrazine hydrochloride (Tetramethylpyrazine Tetrapyrazine) is a chemical compound found in natto and in fermented cocoa beans with anti-inflammation antioxidant antiplatelet and antiapoptosis activities
Catalog No. 50-310-9635 Supplier Selleck Chemical LLC Supplier No. S377525MG
Ligustrazine hydrochloride (Tetramethylpyrazine Tetrapyrazine) is a chemical compound found in natto and in fermented cocoa beans with anti-inflammation antioxidant antiplatelet and antiapoptosis activities