Learn More
Medchemexpress LLC Ligustrazine hydrochloride | 76494-51-4 | 1109103 | 172.66 | 100 MG
SDP
Catalog No. 5000416988
Encompass_Preferred
Ligustrazine (hydrochloride) is a natural product that has shown a protective effect on injured ECV304 cells, significantly increasing NOS and NO formation compared to the model group.
  • Natural product.
  • Protective effect on injured ECV304 cells.
  • Significantly increases NOS and NO formation.
  • Purity: 99.93%.
Catalog No. 50-004-16988 Supplier Medchemexpress LLC Supplier No. HYN0935100MG
Ligustrazine (hydrochloride) is a natural product that has shown a protective effect on injured ECV304 cells, significantly increasing NOS and NO formation compared to the model group.
  • Natural product.
  • Protective effect on injured ECV304 cells.
  • Significantly increases NOS and NO formation.
  • Purity: 99.93%.