Learn More
Medchemexpress LLC Ligustrazine hydrochloride | 76494-51-4 | 25 MG
SDP
Catalog No. 5000389362
Encompass_Preferred
Ligustrazine hydrochloride is a natural product primarily utilized for research. It has shown a protective effect on injured ECV304 cells, significantly increasing NOS and NO formation. This compound is intended for research use only, not for patient or medical applications.
  • Protective effect on injured ECV304 cells
  • Significantly increases NOS and NO formation
  • Suitable for research applications
  • Solid storage at 4°C, sealed and away from moisture
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-89362 Supplier Medchemexpress LLC Supplier No. 25MGHYN0935
Ligustrazine hydrochloride is a natural product primarily utilized for research. It has shown a protective effect on injured ECV304 cells, significantly increasing NOS and NO formation. This compound is intended for research use only, not for patient or medical applications.
  • Protective effect on injured ECV304 cells
  • Significantly increases NOS and NO formation
  • Suitable for research applications
  • Solid storage at 4°C, sealed and away from moisture
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month