Learn More
Cayman Chemical LIH383 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502919354
Encompass_Preferred
A peptide agonist of CXCR7; selectively activates CXCR7 (EC50 = 0.61 nM) over μ-, δ-, and κ-opioid receptors and the nociceptin opioid peptide (NOP) receptor in β-arrestin recruitment assays at 3 µM; prevents dynorphin A-induced uptake of opioid peptides by CXCR7 in U87-ACKR3 cells at 3 µM
Catalog No. 50-291-9354 Supplier Cayman Chemical Supplier No. 3696710 MG
A peptide agonist of CXCR7; selectively activates CXCR7 (EC50 = 0.61 nM) over μ-, δ-, and κ-opioid receptors and the nociceptin opioid peptide (NOP) receptor in β-arrestin recruitment assays at 3 µM; prevents dynorphin A-induced uptake of opioid peptides by CXCR7 in U87-ACKR3 cells at 3 µM