Learn More
Tecan DiTi Box 10/50/200L Ref
Catalog No. NC0775973
Encompass_Preferred
Disposable Tip (DiTi) Box, empty for use with 10/50/200 L refill insert tips, ANSI Format. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. For use with the Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Set of 10.
This item is not returnable. View return policy
Catalog No. NC0775973 Supplier Tecan Supplier No. 30058506
Disposable Tip (DiTi) Box, empty for use with 10/50/200 L refill insert tips, ANSI Format. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. For use with the Freedom EVO Liquid Handling Arm™ (LiHa) or Fluent Flexible Channel Arm™ (FCA). Set of 10.