Learn More
Leica Biosystems LILAC JET ROUTINE III TAPED CA
Supplier:  Leica Biosystems 38440208
Encompass_Preferred
Molded from a special high density acetal polymer, the Leica IP III Tissue Cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of solvents used in histology laboratories.The efficient flow-through slots maximise fluid exchange and ensure proper drainage.
Catalog No. 50-255-2419