Learn More
ACROBiosystems Biotinylated Human LILRA1 Protein, His, Avitag™, 200 μg
Catalog No. 501658067
Encompass_Preferred
Biotinylated human LILRA1, also known as CD85i or LIR-6, is a leukocyte immunoglobulin-like receptor involved in immune regulation. This recombinant protein is expressed in human HEK293 cells and features a C-terminal polyhistidine tag and an Avi tag for site-specific biotinylation.
  • Expressed from human HEK293 cells
  • Site-specific biotinylation via enzymatic labeling
  • Greater than 90% purity as determined by SDS-PAGE
  • Calculated molecular weight of 52.2 kDa, migrating as 60-90 kDa due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose protectant
Catalog No. 50-165-8067 Supplier ACROBiosystems Supplier No. LI1H82E8200UG
Biotinylated human LILRA1, also known as CD85i or LIR-6, is a leukocyte immunoglobulin-like receptor involved in immune regulation. This recombinant protein is expressed in human HEK293 cells and features a C-terminal polyhistidine tag and an Avi tag for site-specific biotinylation.
  • Expressed from human HEK293 cells
  • Site-specific biotinylation via enzymatic labeling
  • Greater than 90% purity as determined by SDS-PAGE
  • Calculated molecular weight of 52.2 kDa, migrating as 60-90 kDa due to glycosylation
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose protectant