Learn More
Aviva Systems Biology Corp LILRA3 Antibody - C-terminal region (ARP65339_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65339_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LILRA3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human LILRA3. Alternate names: CD85E, HM31, HM43, ILT6, LIR-4, LIR4, e3ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-288-478