Learn More
Genscript Corporation LILRA5/CD85f/ILT11Biotin Hi
Supplier:  Genscript Corporation Z04539100
Encompass
The leukocyte immunoglobulin-like receptors (LILR) comprise a family of activating and inhibitory type immunoreceptors whose genes are located in the same locus that encodes killer cell Ig-like receptors (KIRs) Human LILRA5 also known as ILT11 LIR-9 and CD85f consists of a 227 amino acid (aa) extracellular domain (ECD) a 21 aa transmembrane segment and a 10 aa cytoplasmic tail LILRA5 may play a role in triggering innate immune responses Does not seem to play a role for any class I MHC antigen recognition
Catalog No. 50-000-59381