Learn More
Genscript Corporation LILRA6/CD85b/ILT8Biotin His
Catalog No. 5000057203
Encompass
The LILRs are a family of receptors that regulate the activities of myelomonocytic cells Specific allelic variants of two related members of the LILR family LILRB3 and LILRA6 interact with a ligand exposed on necrotic glandular epithelial cells The extracellular domains of LILRB3 and LILRA6 are very similar and their genes are highly polymorphic
Catalog No. 50-000-57203 Supplier Genscript Corporation Supplier No. Z04540100
The LILRs are a family of receptors that regulate the activities of myelomonocytic cells Specific allelic variants of two related members of the LILR family LILRB3 and LILRA6 interact with a ligand exposed on necrotic glandular epithelial cells The extracellular domains of LILRB3 and LILRA6 are very similar and their genes are highly polymorphic