Learn More
Sino Biological Recombinant Human ILT-8/LILRA6 Protein (hFc Tag)
Supplier:  Sino Biological 29813H02H100
Encompass_Preferred
A DNA sequence encoding the human LILRA6 (Q6PI73-1) (Met1-Asn447) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-6453