Learn More
ACROBiosystems Recombinant Protein;Human LILRB1 / CD85j / ILT2 Protein, Fc Tag;HEK293;100UG;CDJ,CD85J,LILRB1,CD85,ILT-2,ILT2,LIR-1,LIR1,MIR-7,MIR7,LIR1
Catalog No. 501658154
Encompass_Preferred
Recombinant Protein;Human LILRB1 / CD85j / ILT2 Protein, Fc Tag;100UG;Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # NP_006660).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8154 Supplier ACROBiosystems Supplier No. CDJH5252100UG
Recombinant Protein;Human LILRB1 / CD85j / ILT2 Protein, Fc Tag;100UG;Human LILRB1, Fc Tag (CDJ-H5252) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # NP_006660).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 73.8 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.