Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LILRB1 / CD85j / ILT2 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CDJ,CD85J,LILRB1,CD85,ILT-2,ILT2,LIR-1,LIR1,MIR-7,MIR7,LIR1
Catalog No. 501658367
Encompass_Preferred
Recombinant Protein;Biotinylated Human LILRB1 / CD85j / ILT2 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRB1, Fc Tag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM5-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.6 kDa. As a result of glycosylation, the protein migrates as 100-116 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8367 Supplier ACROBiosystems Supplier No. CDJH82F725UG
Recombinant Protein;Biotinylated Human LILRB1 / CD85j / ILT2 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRB1, Fc Tag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM5-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.6 kDa. As a result of glycosylation, the protein migrates as 100-116 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).