Learn More
Aviva Systems Biology Corp LILRB2 Antibody - C-terminal region (ARP63917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LILRB2. It was validated on Western Blot. . Alternate names: CD85D, ILT4, LILRA6, LIR-2, LIR2, MIR-10, MIR10ApplicationWB. Species reactivity: Human
Catalog No. 50-288-116