Learn More
Genscript Corporation LILRB2/CD85d/ILT4Biotin His
Catalog No. 5000059295
Encompass
The immunoglobulin-like transcript (ILT) comprise a family of activating and inhibitory type immunoreceptors whose genes are located in the same locus that encodes killer cell Ig-like receptors (KIR) ILT4 also known as LIR-2 and LILRB2 is a type I transmembrane protein expressed primarily on monocytes and dendritic cells (DC) LILRB2 is a receptor for class I MHC antigens Recognizes a broad spectrum of HLA-A HLA-B HLA-C HLA-G and HLA-F alleles
Catalog No. 50-000-59295 Supplier Genscript Corporation Supplier No. Z04542500
The immunoglobulin-like transcript (ILT) comprise a family of activating and inhibitory type immunoreceptors whose genes are located in the same locus that encodes killer cell Ig-like receptors (KIR) ILT4 also known as LIR-2 and LILRB2 is a type I transmembrane protein expressed primarily on monocytes and dendritic cells (DC) LILRB2 is a receptor for class I MHC antigens Recognizes a broad spectrum of HLA-A HLA-B HLA-C HLA-G and HLA-F alleles