Learn More
ACROBiosystems Recombinant Protein;Human LILRB3 / CD85a / ILT5 Protein, Fc Tag;HEK293;100UG;CDA,LILRB3,CD85a,ILT5,LIR-3,ILT-5
Supplier: ACROBiosystems CDAH5250100UG
Recombinant Protein;Human LILRB3 / CD85a / ILT5 Protein, Fc Tag;100UG;Human LILRB3, Fc Tag (CDA-H5250) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAB68668(P288R)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.8 kDa. As a result of glycosylation, the protein migrates as 90-115 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).