Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LILRB3 / CD85a / ILT5 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CDA,LILRB3,CD85a,ILT5,LIR-3,ILT-5
Supplier:  ACROBiosystems CDAH82F625UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LILRB3 / CD85a / ILT5 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRB3, Fc Tag (CDA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAI12199).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.5 kDa. As a result of glycosylation, the protein migrates as 80-105 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8365