Learn More
Biomatik Corporation Lilrb4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10728
Encompass_Preferred
Lilrb4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LILRB4. The Lilrb4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Leukocyte Immunoglobulin-Like Receptor Subfamily B Member 4 Protein (22-259Aa). Applications: ELISA, WB, IHC. Target Synonyms: LILRB4. Accession Number: Q8NHJ6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9261