Learn More
Genscript Corporation LILRB4/CD85k/ILT3Biotin His
Supplier:  Genscript Corporation Z04546500
Encompass
LILRB4 also known as CD85k and LIR-5 ILT3 is an approximately 60 kDa transmembrane glycoprotein that negatively regulates immune cell activation Mature human ILT3 consists of a 238 amino acid (aa) extracellular domain with two Ig-like domains a 21 aa transmembrane segment and a 168 aa cytoplasmic domain with 3 immunoreceptor tyrosine-based inhibitory motifs (ITIM) LILRB4 is receptor for class I MHC antigens Recognizes a broad spectrum of HLA-A HLA-B HLA-C and HLA-G alleles
Catalog No. 50-000-57221