Learn More
ACROBiosystems ACROBiosystems Cynomolgus LILRB4 / CD85k / ILT3 Protein, His Tag
Catalog No. 501957276
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus LILRB4, His Tag (CDK-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LILRB4, His Tag (Cat. No. CDK-C5227) at 2 μg/mL (100 μL/well) can bind Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) with a linear range of 8-125 ng/mL (QC tested).
Catalog No. 50-195-7276 Supplier ACROBiosystems Supplier No. CDKC52271MG
Recombinant Protein;1MG;Cynomolgus LILRB4, His Tag (CDK-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LILRB4, His Tag (Cat. No. CDK-C5227) at 2 μg/mL (100 μL/well) can bind Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) with a linear range of 8-125 ng/mL (QC tested).