Learn More
ACROBiosystems ACROBiosystems Human LILRB4 / CD85k / ILT3 Protein, His Tag
Supplier:  ACROBiosystems LI4H52H71MG
Encompass_Preferred
Recombinant Protein;1MG;Human LILRB4, His Tag (LI4-H52H7) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.2 kDa.
Catalog No. 50-195-6768