Learn More
Sino Biological LILRB4/CD85k/ILT3 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 16742MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LILRB4/CD85k/ILT3 (rh LILRB4/CD85k/ILT3; Catalog#16742-H08H; NP_001265357.2; Met1-Glu259). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8411