Learn More
Genscript Corporation LILRB5/CD85c/LIR-8Biotin Hi
Catalog No. 5000059418
Encompass
LILRB5 also known as CD85c andLIR-8 belongs to a family of transmembrane glycoproteins that negatively regulate immune cell activation Mature human LIR-8 consists of a 435 amino acid (aa) extracellular domain with four Ig-like domains a 21 aa transmembrane segment and a 111 aa cytoplasmic domain with two immunoreceptor tyrosine-based inhibitory motifs (ITIM) LILRB5 may act as receptor for class I MHC antigens
Catalog No. 50-000-59418 Supplier Genscript Corporation Supplier No. Z04548500
LILRB5 also known as CD85c andLIR-8 belongs to a family of transmembrane glycoproteins that negatively regulate immune cell activation Mature human LIR-8 consists of a 435 amino acid (aa) extracellular domain with four Ig-like domains a 21 aa transmembrane segment and a 111 aa cytoplasmic domain with two immunoreceptor tyrosine-based inhibitory motifs (ITIM) LILRB5 may act as receptor for class I MHC antigens