Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LILRB5 / CD85c / LIR-8 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;CDC,LILRB5,CD85c,LIR-8,LIR8
Supplier:  ACROBiosystems CDCH82F8200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LILRB5 / CD85c / LIR-8 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human LILRB5, Fc Tag (CDC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # O75023-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.8 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRB5, Fc Tag (Cat. No. CDC-H82F8) with a linear range of 0.156-5 μg/mL (QC tested).
Catalog No. 50-165-8404