Learn More
Cusabio Technology LLC LIM and SH3 protein 1
Catalog No. 5000051866
Encompass_Preferred
This kit detects LASP1 in human serum plasma tissue homogenates and cell lysates employing a quantitative sandwich ELISA method With a detection range of 25 pg/mL to 1600 pg/mL and a sensitivity of 6 25 pg/mL it enables precise measurement of LASP1 concentrations across diverse biological samples The assay requires 50-100 L of sample per test and can be completed within 1 to 5 hours depending on protocol optimization Absorbance readings are taken at 450 nm ensuring reliable and reproducible results for research applications requiring accurate quantification of LASP1 protein levels in clinical or experimental settings
Catalog No. 50-000-51866 Supplier Cusabio Technology LLC Supplier No. CSBEL012760HU
This kit detects LASP1 in human serum plasma tissue homogenates and cell lysates employing a quantitative sandwich ELISA method With a detection range of 25 pg/mL to 1600 pg/mL and a sensitivity of 6 25 pg/mL it enables precise measurement of LASP1 concentrations across diverse biological samples The assay requires 50-100 L of sample per test and can be completed within 1 to 5 hours depending on protocol optimization Absorbance readings are taken at 450 nm ensuring reliable and reproducible results for research applications requiring accurate quantification of LASP1 protein levels in clinical or experimental settings