Learn More
Aviva Systems Biology Corp LIM2 Antibody - N-terminal region (ARP49954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LIM2. Alternate names: MP19, MP17ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60225