Learn More
Aviva Systems Biology Corp LIMCH1 Antibody - C-terminal region (ARP62677_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62677_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIMCH1. It was validated on Western Blot. . Alternate names: DKFZp434I0312, DKFZp686A01247, DKFZp686B2470, DKFZp686G18243, DKFZp686G2094, DKFZp781C1754, DKFZp781I1455, LIMCH1A, LMO7B, MGC72127ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-735