Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lime Water
SCI_ED

Catalog No. S25386A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
3.785 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25386A 3.785 L
S25386 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25386A
Detection of CO2

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Lime Water
Color Colorless
CAS 1305-62-0, 7732-18-5
Assay Percent Range 14%
Density 1g/mL
Packaging Solution
Quantity 3.785 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.