Learn More
Grainger Limit Switch Body 2NO/2NC
Catalog No. 502750690
Encompass_Preferred
The Dayton Limit Switch Body is a limit switch with 2 normally open and 2 normally closed contacts. It has a NEMA 1 general purpose enclosure rating and is suitable for indoor use. The operating temperature range is -20°F to 140°F. The electrical rating is 10A at 125/250VAC and 1/4HP at 125VAC. The switch actuates via a plunger with 0.35" overtravel. The body is made of die cast zinc alloy and has epoxy coating. It can be mounted in any orientation. The switch body dimensions are 1.89" x 1.26" x 2.36". It weighs 0.48 lbs.
Catalog No. 50-275-0690 Supplier Grainger Supplier No. 14H178
The Dayton Limit Switch Body is a limit switch with 2 normally open and 2 normally closed contacts. It has a NEMA 1 general purpose enclosure rating and is suitable for indoor use. The operating temperature range is -20°F to 140°F. The electrical rating is 10A at 125/250VAC and 1/4HP at 125VAC. The switch actuates via a plunger with 0.35" overtravel. The body is made of die cast zinc alloy and has epoxy coating. It can be mounted in any orientation. The switch body dimensions are 1.89" x 1.26" x 2.36". It weighs 0.48 lbs.