Learn More
Grainger Limit Switch Box
Catalog No. 502752509
Encompass_Preferred
The Approved Vendor Limit Switch Box is a heavy-duty NEMA 4X rated enclosure for harsh environments. It features 14 gauge steel construction, continuous weld seams, and stainless steel hardware for durability. The powder-coated enclosure measures 8 x 6 x 4 inches and has a transparent shatter-resistant polycarbonate cover with four stainless steel quarter-turn latches for easy access. It protects contents from dirt, rain, sleet, snow, and water, making it ideal for indoor and outdoor industrial applications, ensuring reliable protection for limit switch equipment.
Catalog No. 50-275-2509 Supplier Grainger Supplier No. 14N260
The Approved Vendor Limit Switch Box is a heavy-duty NEMA 4X rated enclosure for harsh environments. It features 14 gauge steel construction, continuous weld seams, and stainless steel hardware for durability. The powder-coated enclosure measures 8 x 6 x 4 inches and has a transparent shatter-resistant polycarbonate cover with four stainless steel quarter-turn latches for easy access. It protects contents from dirt, rain, sleet, snow, and water, making it ideal for indoor and outdoor industrial applications, ensuring reliable protection for limit switch equipment.