Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-Limonene

Catalog No. 621221LF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-122-1LF 1 L
62-122-250MLF 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 62-122-1LF Supplier Sigma Aldrich Supplier No. 621221LF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-Limonene is present in the peel oil from citrus fruits. It is the most commonly used fragrance materials in technical products and in fine fragrances. It is the major constituent of the essential oil extracted from Anethum sowa Roxb. (Indian dill). Racemic limonene undergoes enantio-selective and enantio-specific conversion to (R)-(+)-+a-terpineol byPenicillium digitatum mycelia. Biotransformation of (R)-(+)- and (S)-(-)-limonene by fungi has been investigated.(R)-(+)-Limonene has been used to investigate the in vitro and in vivo action of limonene against Leishmania species.

Specifications

CAS 5989-27-5
Density 0.842 g/mL at 20°C
Boiling Point 176°C to 177°C
Molecular Formula C10H16
Refractive Index n20/D 1.473
Linear Formula C10H16
MDL Number MFCD00062991
Quantity 1 L
Synonym (+)-p-Mentha-1,8-diene; (+)-Carvene; (R)-4-Isopropenyl-1-methyl-1-cyclohexene
Molecular Weight (g/mol) 136.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.