Learn More
Electron Microscopy Sciences Limonene-Mount (Substitute for BioMeda Clarion Mount) 100 ML
SDP
Supplier:  Electron Microscopy Sciences 1798701
Encompass_Preferred
This mounting medium is made with limonene, a natural product from orange peels. It preserves  tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt. Our Limonene-Mount also works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red. It is also a good choice for mounting H and E stained slides. Application: Mounting tissues and cell smears.
Catalog No. 50-302-44