Learn More
Electron Microscopy Sciences Limonene-Mount (Substitute for BioMeda Clarion Mount) 250 ML
SDP
Catalog No. 5030245
Encompass_Preferred
This mounting medium is made with limonene, a natural product from orange peels. It preserves  tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt. Our Limonene-Mount also works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red. It is also a good choice for mounting H and E stained slides. Application: Mounting tissues and cell smears.
Catalog No. 50-302-45 Supplier Electron Microscopy Sciences Supplier No. 1798725
This mounting medium is made with limonene, a natural product from orange peels. It preserves  tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt. Our Limonene-Mount also works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red. It is also a good choice for mounting H and E stained slides. Application: Mounting tissues and cell smears.