Learn More
Abcam Limonene Mounting Medium, 30ML
Catalog No. 502585552
Encompass_Preferred
Limonene Mounting Medium ab104141 is an organic mounting medium compatible with chromogens such as DAB.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5552 Supplier Abcam Supplier No. AB10414130ML
Limonene Mounting Medium ab104141 is an organic mounting medium compatible with chromogens such as DAB.
The product is subject to the following: Abcam Restricted Use Statement