Learn More
Medchemexpress LLC Limonin | 1180-71-4 | >98% | 470.51 | 1 ML
SDP
Catalog No. 5000385961
Encompass_Preferred
Limonin is a natural product isolated from Dictamnus dasycarpus. It is known for its antiviral and antitumor activities, inhibiting HIV-1 and inducing apoptosis in human colon adenocarcinoma cells. In vitro studies show limonin's anti-proliferation effects on colon adenocarcinoma cells and pancreatic cancer cell lines, while in vivo research demonstrates its ability to significantly inhibit tumor growth in xenograft mouse models.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM.
  • Shows anti-proliferation effects on Caco-2 and HCT-116 cells.
  • Suppresses cell migration and invasion in HCT-116 cells.
  • Activates Erk and p38 MAPK pathways.
  • Significantly inhibits tumor growth in HCT-116 xenograft mouse models.
Catalog No. 50-003-85961 Supplier Medchemexpress LLC Supplier No. HY1741110MM/1ML
Limonin is a natural product isolated from Dictamnus dasycarpus. It is known for its antiviral and antitumor activities, inhibiting HIV-1 and inducing apoptosis in human colon adenocarcinoma cells. In vitro studies show limonin's anti-proliferation effects on colon adenocarcinoma cells and pancreatic cancer cell lines, while in vivo research demonstrates its ability to significantly inhibit tumor growth in xenograft mouse models.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM.
  • Shows anti-proliferation effects on Caco-2 and HCT-116 cells.
  • Suppresses cell migration and invasion in HCT-116 cells.
  • Activates Erk and p38 MAPK pathways.
  • Significantly inhibits tumor growth in HCT-116 xenograft mouse models.