Learn More
Apexbio Technology LLC Limonin 1180-71-8 200mg
SDP
Catalog No. 5000093699
Encompass_Preferred
Limonin (CAS 1180-71-8) is a natural limonoid with inhibitory activities against carcinogenesis and HIV-1 replication It modulates apoptotic signaling pathways to induce apoptosis and suppress HIV-1 replication In in vitro studies limonin demonstrates dose-dependent inhibition of HIV-1 replication with an EC50 of approximately 60 M in human peripheral blood mononuclear cells (PBMCs) and monocyte/macrophage (M/M) cells In human hepatoma HepG2 cells limonin induces apoptosis by affecting relevant signaling pathways Animal studies show that limonin reduces tumor burden and incidence in DMBA-treated Syrian hamsters and inhibits colon adenocarcinoma development in azoxymethane (AOM)-treated rats Based on these pharmacological properties limonin holds research potential in HIV antiviral mechanisms and chemically-induced carcinogenesis models
Catalog No. 50-000-93699 Supplier Apexbio Technology LLC Supplier No. A3555200
Limonin (CAS 1180-71-8) is a natural limonoid with inhibitory activities against carcinogenesis and HIV-1 replication It modulates apoptotic signaling pathways to induce apoptosis and suppress HIV-1 replication In in vitro studies limonin demonstrates dose-dependent inhibition of HIV-1 replication with an EC50 of approximately 60 M in human peripheral blood mononuclear cells (PBMCs) and monocyte/macrophage (M/M) cells In human hepatoma HepG2 cells limonin induces apoptosis by affecting relevant signaling pathways Animal studies show that limonin reduces tumor burden and incidence in DMBA-treated Syrian hamsters and inhibits colon adenocarcinoma development in azoxymethane (AOM)-treated rats Based on these pharmacological properties limonin holds research potential in HIV antiviral mechanisms and chemically-induced carcinogenesis models