Learn More
Medchemexpress LLC Limonin | 1180-71-8 | 99.4% | 470.51 | 100 MG
SDP
Catalog No. 5000384269
Encompass_Preferred
Limonin inhibits HIV-1 with an EC50 of 60.0 μM. It induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM. Limonin possesses antiviral and antitumor activities, making it relevant for research in infection and cancer.
  • Inhibits HIV-1.
  • Induces apoptosis in human colon adenocarcinoma cells.
  • Exhibits antiviral and antitumor activities.
  • Targets HIV.
  • Targets apoptosis.
  • Targets endogenous metabolite.
  • Targets cytochrome P450.
  • Targets P-glycoprotein.
Catalog No. 50-003-84269 Supplier Medchemexpress LLC Supplier No. HY17411100MG
Limonin inhibits HIV-1 with an EC50 of 60.0 μM. It induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM. Limonin possesses antiviral and antitumor activities, making it relevant for research in infection and cancer.
  • Inhibits HIV-1.
  • Induces apoptosis in human colon adenocarcinoma cells.
  • Exhibits antiviral and antitumor activities.
  • Targets HIV.
  • Targets apoptosis.
  • Targets endogenous metabolite.
  • Targets cytochrome P450.
  • Targets P-glycoprotein.