Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Limonin | 1180-71-8 | MFCD00075922 | 99.1% | 470.51 g/mol | C26H30O8 | 10 MG
SDP

Catalog No. 5000299173
Encompass_Preferred

Limonin (limonoic acid 3,19:16,17 dilactone) is provided as an analytical reference standard intended for research and analytical applications. This small-quantity reference material is used for assay development, method validation, and quantitative analysis where a traceable, high-purity standard is required.

  • Analytical reference standard of limonin (limonoic acid 3,19:16,17 dilactone).
  • Purity 99.07% as listed on the product page.
  • CAS number 1180-71-8.
  • Molecular weight 470.51 g/mol.
  • Suitable for assay development, quantification, and method validation.
  • Available in small package sizes intended for analytical use.

Catalog No. 50-002-99173 Supplier Medchemexpress LLC Supplier No. HY17411R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Limonin (limonoic acid 3,19:16,17 dilactone) is provided as an analytical reference standard intended for research and analytical applications. This small-quantity reference material is used for assay development, method validation, and quantitative analysis where a traceable, high-purity standard is required.

  • Analytical reference standard of limonin (limonoic acid 3,19:16,17 dilactone).
  • Purity 99.07% as listed on the product page.
  • CAS number 1180-71-8.
  • Molecular weight 470.51 g/mol.
  • Suitable for assay development, quantification, and method validation.
  • Available in small package sizes intended for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.