Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Limonin

Catalog No. AC465472500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465472500 250 mg
AC465470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465472500 Supplier Thermo Scientific Chemicals Supplier No. 465472500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1180-71-8 | C26H30O8 | 470.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1180-71-8
Molecular Weight (g/mol) 470.52
MDL Number MFCD00075922
InChI Key KBDSLGBFQAGHBE-MSGMIQHVSA-N
IUPAC Name (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
SMILES CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1

Specifications

Alpha Vector LIMONIN
Melting Point 298°C to 300°C
Color White
Loss on Drying ≤5 %
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C26H30O8
Quantity 250 mg
Formula Weight 470.52
Physical Form Powder
Chemical Name or Material Limonin
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.